ContactUsLinkComponent
Mouse 68kDa Neurofilament/NF-L ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of mouse 68kDa Neurofilament/NF-L in serum, plasma, cerebrospinal fluid or other biological fluids 1 * 48 Tests
Catalog # ANTIA6516-48
Supplier:  Antibodies.com
New Product
Mouse 68kDa Neurofilament/NF-L ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of mouse 68kDa Neurofilament/NF-L in serum, plasma, cerebrospinal fluid or other biological fluids 1 * 48 Tests
Catalog # ANTIA6516-48
Supplier:  Antibodies.com