ContactUsLinkComponent
Human SAHH ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human SAHH in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6737-48
Supplier:  Antibodies.com
New Product
Human SAHH ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human SAHH in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6737-48
Supplier:  Antibodies.com