ContactUsLinkComponent
Human Deoxyribonuclease I Like 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of human Deoxyribonuclease I Like 1 in tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA8025-48
New Product
Human Deoxyribonuclease I Like 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of human Deoxyribonuclease I Like 1 in tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA8025-48