ContactUsLinkComponent
Rat beta 3 Adrenergic Receptor ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of rat beta 3 Adrenergic Receptor in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7266-48
Supplier:  ANTIBODIES.COM
New Product
Rat beta 3 Adrenergic Receptor ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of rat beta 3 Adrenergic Receptor in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7266-48
Supplier:  ANTIBODIES.COM