ContactUsLinkComponent
Bovine Orexin A ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determi nation of bovine Orexin A in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA82335-48
Supplier:  Antibodies.com
New Product
Bovine Orexin A ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determi nation of bovine Orexin A in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA82335-48
Supplier:  Antibodies.com