ContactUsLinkComponent
Porcine HGF ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of porcine HGF in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA82280-48
Supplier:  ANTIBODIES.COM
New Product
Porcine HGF ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of porcine HGF in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA82280-48
Supplier:  ANTIBODIES.COM