Human Antithrombin III/ATIII ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitati ve determination of human Antithrombin III/ATIII in serum, plasma, tissue homogenates, cell lysates, cell culture sup 1 * 48 Tests
Catalog # ANTIA6321-48
Supplier: Antibodies.com