ContactUsLinkComponent
Rat cAMP Protein Kinase Catalytic Subunit alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat cAMP Protein Kinase Catalytic Subunit alpha in serum, plasma, tissue homoge 1 * 96 Tests
Catalog # ANTIA78667-96
Supplier:  ANTIBODIES.COM
New Product
Rat cAMP Protein Kinase Catalytic Subunit alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat cAMP Protein Kinase Catalytic Subunit alpha in serum, plasma, tissue homoge 1 * 96 Tests
Catalog # ANTIA78667-96
Supplier:  ANTIBODIES.COM