ContactUsLinkComponent
Human Smooth Muscle Myosin Heavy Chain 11 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of human Smooth Muscle Myosin Heavy Chain 11 in serum, plasma, tissue homogenates, and 1 * 96 Tests
Catalog # ANTIA77000-96
Supplier:  ANTIBODIES.COM
New Product
Human Smooth Muscle Myosin Heavy Chain 11 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of human Smooth Muscle Myosin Heavy Chain 11 in serum, plasma, tissue homogenates, and 1 * 96 Tests
Catalog # ANTIA77000-96
Supplier:  ANTIBODIES.COM