ContactUsLinkComponent
Human MA2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human MA2 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7442-48
Supplier:  ANTIBODIES.COM
New Product
Human MA2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human MA2 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7442-48
Supplier:  ANTIBODIES.COM