ContactUsLinkComponent
Mouse DHODH ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse DHODH in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6916-48
Supplier:  ANTIBODIES.COM
New Product
Mouse DHODH ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse DHODH in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6916-48
Supplier:  ANTIBODIES.COM