ContactUsLinkComponent
Human Filamin A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human Filamin A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologica 1 * 48 Tests
Catalog # ANTIA7570-48
Supplier:  ANTIBODIES.COM
New Product
Human Filamin A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human Filamin A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologica 1 * 48 Tests
Catalog # ANTIA7570-48
Supplier:  ANTIBODIES.COM