ContactUsLinkComponent
Chicken Glucagon Like Peptide 1 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quan titative determination of chicken Glucagon Like Peptide 1 in serum, plasma, tissue homogenates, cell lysates, cell cu 1 * 48 Tests
Catalog # ANTIA82412-48
Supplier:  ANTIBODIES.COM
New Product
Chicken Glucagon Like Peptide 1 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quan titative determination of chicken Glucagon Like Peptide 1 in serum, plasma, tissue homogenates, cell lysates, cell cu 1 * 48 Tests
Catalog # ANTIA82412-48
Supplier:  ANTIBODIES.COM