Human Hepatitis B Virus E Antigen Antibody ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro qualitative determination of human Hepatitis B Virus E Antigen Antibody in serum, plasma, tissue homogenates, s 1 * 96 Tests
Catalog # ANTIA76055-96
Supplier: ANTIBODIES.COM