ContactUsLinkComponent
Canine Transferrin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of canine Transferrin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6960-48
Supplier:  ANTIBODIES.COM
New Product
Canine Transferrin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of canine Transferrin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6960-48
Supplier:  ANTIBODIES.COM