ContactUsLinkComponent
Human Tbx3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human Tbx3 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6442-48
Supplier:  ANTIBODIES.COM
New Product
Human Tbx3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human Tbx3 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6442-48
Supplier:  ANTIBODIES.COM