ContactUsLinkComponent
Human TGF beta 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human TGF beta 2 in serum, plasma, tissue homogenates, cell culture supernates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA8116-96
Supplier:  ANTIBODIES.COM
New Product
Human TGF beta 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human TGF beta 2 in serum, plasma, tissue homogenates, cell culture supernates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA8116-96
Supplier:  ANTIBODIES.COM