ContactUsLinkComponent
Human Defensin beta 126 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of human Defensin beta 126 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or 1 * 96 Tests
Catalog # ANTIA6590-96
Supplier:  ANTIBODIES.COM
New Product
Human Defensin beta 126 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of human Defensin beta 126 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or 1 * 96 Tests
Catalog # ANTIA6590-96
Supplier:  ANTIBODIES.COM