ContactUsLinkComponent
Human Glutathione Synthetase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitati ve determination of human Glutathione Synthetase in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6678-48
Supplier:  ANTIBODIES.COM
New Product
Human Glutathione Synthetase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitati ve determination of human Glutathione Synthetase in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6678-48
Supplier:  ANTIBODIES.COM