ContactUsLinkComponent
Human TNFRSF1B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human TNFRSF1B in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6915-48
Supplier:  ANTIBODIES.COM
New Product
Human TNFRSF1B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human TNFRSF1B in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6915-48
Supplier:  ANTIBODIES.COM