ContactUsLinkComponent
Human Nicotinic Acetylcholine Receptor alpha 5/CHRNA5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed fo r the in vitro quantitative determination of human Nicotinic Acetylcholine Receptor alpha 5/CHRNA5 in tissue homogena 1 * 48 Tests
Catalog # ANTIA5971-48
Supplier:  Antibodies.com
New Product
Human Nicotinic Acetylcholine Receptor alpha 5/CHRNA5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed fo r the in vitro quantitative determination of human Nicotinic Acetylcholine Receptor alpha 5/CHRNA5 in tissue homogena 1 * 48 Tests
Catalog # ANTIA5971-48
Supplier:  Antibodies.com