ContactUsLinkComponent
Guinea Pig Gastrin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of guinea pig Gastrin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7839-48
Supplier:  Antibodies.com
New Product
Guinea Pig Gastrin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of guinea pig Gastrin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7839-48
Supplier:  Antibodies.com