ContactUsLinkComponent
Human MCHR-1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human MCHR-1 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7006-48
Supplier:  ANTIBODIES.COM
New Product
Human MCHR-1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human MCHR-1 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7006-48
Supplier:  ANTIBODIES.COM