ContactUsLinkComponent
Mouse Synapsin I ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of mouse Synapsin I in tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA8072-48
Supplier:  ANTIBODIES.COM
New Product
Mouse Synapsin I ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of mouse Synapsin I in tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA8072-48
Supplier:  ANTIBODIES.COM