ContactUsLinkComponent
Human Gli1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human Gli1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Katalogové číslo ANTIA7050-48
Supplier:  Antibodies.com
New Product
Human Gli1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human Gli1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Katalogové číslo ANTIA7050-48
Supplier:  Antibodies.com