ContactUsLinkComponent
Human RFK ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human RFK in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6754-48
Supplier:  ANTIBODIES.COM
New Product
Human RFK ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human RFK in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6754-48
Supplier:  ANTIBODIES.COM