ContactUsLinkComponent
Human PARN ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human PARN in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6242-48
Supplier:  ANTIBODIES.COM
New Product
Human PARN ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human PARN in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6242-48
Supplier:  ANTIBODIES.COM