ContactUsLinkComponent
Human Liver Carboxylesterase 1/CES1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of human Liver Carboxylesterase 1/CES1 in serum, plasma, tissue homogenates or other biologic 1 * 48 Tests
Catalog # ANTIA82313-48
Supplier:  ANTIBODIES.COM
New Product
Human Liver Carboxylesterase 1/CES1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of human Liver Carboxylesterase 1/CES1 in serum, plasma, tissue homogenates or other biologic 1 * 48 Tests
Catalog # ANTIA82313-48
Supplier:  ANTIBODIES.COM