ContactUsLinkComponent
Human WFS1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human WFS1 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6382-48
Supplier:  Antibodies.com
New Product
Human WFS1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human WFS1 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6382-48
Supplier:  Antibodies.com