ContactUsLinkComponent
Human Casein kappa ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of human Casein kappa in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA8101-48
Supplier:  ANTIBODIES.COM
New Product
Human Casein kappa ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of human Casein kappa in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA8101-48
Supplier:  ANTIBODIES.COM