ContactUsLinkComponent
Human PI 3 Kinase Catalytic Subunit alpha/PIK3CA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human PI 3 Kinase Catalytic Subunit alpha/PIK3CA in serum, plasma, tissue hom 1 * 96 Tests
Catalog # ANTIA75722-96
Supplier:  ANTIBODIES.COM
New Product
Human PI 3 Kinase Catalytic Subunit alpha/PIK3CA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human PI 3 Kinase Catalytic Subunit alpha/PIK3CA in serum, plasma, tissue hom 1 * 96 Tests
Catalog # ANTIA75722-96
Supplier:  ANTIBODIES.COM