ContactUsLinkComponent
Human TLS/FUS ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human TLS/FUS in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6627-96
Supplier:  ANTIBODIES.COM
New Product
Human TLS/FUS ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human TLS/FUS in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6627-96
Supplier:  ANTIBODIES.COM