ContactUsLinkComponent
Single use Temperature logger S1 V2 , temperature range : -30 °C to + 70 °C , accuracy : +/- 0,5 °C , resolution : 0,1 °C, start delay : 30 minutes, // Low alarm at 2 °C , High Alarm at 8 °C , No alarm delay 1 * 20 items
Catalog # IMECS1C30A69010
Supplier:  tempmate
New Product
Single use Temperature logger S1 V2 , temperature range : -30 °C to + 70 °C , accuracy : +/- 0,5 °C , resolution : 0,1 °C, start delay : 30 minutes, // Low alarm at 2 °C , High Alarm at 8 °C , No alarm delay 1 * 20 items
Catalog # IMECS1C30A69010
Supplier:  tempmate