ContactUsLinkComponent
Deuterated PAH Mixture - 7 components: 200ug/ml each of Acenaphthene D10 [CAS:15067-26-2] Benzo(a)anthracene D12 [CAS:1718-53-2] Benzo(a)pyrene D12 [CAS:63466-71-7] Dibenzo(a,h)anthracene D14 [CAS:13250-98-1] Dibenzo(a,i)pyrene-d14 [CAS:15877 1 * 1 mL
Catalog # ACSDF872151
Supplier:  CPACHEM
New Product
Deuterated PAH Mixture - 7 components: 200ug/ml each of Acenaphthene D10 [CAS:15067-26-2] Benzo(a)anthracene D12 [CAS:1718-53-2] Benzo(a)pyrene D12 [CAS:63466-71-7] Dibenzo(a,h)anthracene D14 [CAS:13250-98-1] Dibenzo(a,i)pyrene-d14 [CAS:15877 1 * 1 mL
Catalog # ACSDF872151
Supplier:  CPACHEM