ContactUsLinkComponent
Recalibration Calibration Standard UV80 calibration standard for testing the photometric accuracy (1,0 Abs) in the UV range, cleaned and recalibrated at: wavelengths: 235, 257, 313, 350nm slit width: 2.0nm serial number: with DAkkS calibration certif 1 * 1 items
Catalog # HELL667080RE
Supplier:  HELLMA
New Product
Recalibration Calibration Standard UV80 calibration standard for testing the photometric accuracy (1,0 Abs) in the UV range, cleaned and recalibrated at: wavelengths: 235, 257, 313, 350nm slit width: 2.0nm serial number: with DAkkS calibration certif 1 * 1 items
Catalog # HELL667080RE
Supplier:  HELLMA