ContactUsLinkComponent
14 components: Al 500mg/l V 250mg/l As 100mg/l Be 100mg/l Co 100mg/l Cr 100mg/l Cu 100mg/l Fe 100mg/l Mn 100mg/l Ni 100mg/l Pb 100mg/l Zn 100mg/l Cd 25mg/l Se 25mg/l in HNO3 5% ,, CRM, ISO 17034 and ISO 17025 Conservation 12 1 * 125 mL
Catalog # CMPLRM002158L125
New Product
14 components: Al 500mg/l V 250mg/l As 100mg/l Be 100mg/l Co 100mg/l Cr 100mg/l Cu 100mg/l Fe 100mg/l Mn 100mg/l Ni 100mg/l Pb 100mg/l Zn 100mg/l Cd 25mg/l Se 25mg/l in HNO3 5% ,, CRM, ISO 17034 and ISO 17025 Conservation 12 1 * 125 mL
Catalog # CMPLRM002158L125