ContactUsLinkComponent
17 components: 100mg/l each of Benzene [CAS:71-43-2] Bromodichloromethane [CAS:75-27-4] Dibromochloromethane [CAS:124-48-1] Tribromomethane [CAS:75-25-2] Chloroform [CAS:67-66-3] Tetrachloromethane [CAS:56-23-5] 1,1-Dichloroethene [CAS:75 1 * 1 mL
Catalog # CMPLRD0070481.5
New Product
17 components: 100mg/l each of Benzene [CAS:71-43-2] Bromodichloromethane [CAS:75-27-4] Dibromochloromethane [CAS:124-48-1] Tribromomethane [CAS:75-25-2] Chloroform [CAS:67-66-3] Tetrachloromethane [CAS:56-23-5] 1,1-Dichloroethene [CAS:75 1 * 1 mL
Catalog # CMPLRD0070481.5