ContactUsLinkComponent
,,KIT OF 23 components: 1000mg/l each of Dibutyl Disulfide [CAS:629-45-8] Allyl disulfide [CAS:2179-57-9] Diallylsulfide [CAS:592-88-1] n-Butyl Mercaptan [CAS:109-79-5] Dipropyl disulfide [CAS:629-19-6] Diethylsulfide [CAS:352-93-2] Dimet 1 * 1 mL
Catalog # CMPLRD1194261.5
New Product
,,KIT OF 23 components: 1000mg/l each of Dibutyl Disulfide [CAS:629-45-8] Allyl disulfide [CAS:2179-57-9] Diallylsulfide [CAS:592-88-1] n-Butyl Mercaptan [CAS:109-79-5] Dipropyl disulfide [CAS:629-19-6] Diethylsulfide [CAS:352-93-2] Dimet 1 * 1 mL
Catalog # CMPLRD1194261.5