ContactUsLinkComponent
8 components: 50mg/l each of 2-(3 ,5-Di-tert-butyl-2-hydroxyphenyl)-2H-benzotriazole [CAS:3846-71-7] 2 ,4-Di-tert-butyl-6-(5-chloro-2H-benzotriazol-2-yl)phenol [CAS:3864-99-1] 2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol [CAS:25973-55-1] 2 1 * 1 mL
Catalog # ACSDRD1190551.2
Supplier:  CPACHEM
New Product
8 components: 50mg/l each of 2-(3 ,5-Di-tert-butyl-2-hydroxyphenyl)-2H-benzotriazole [CAS:3846-71-7] 2 ,4-Di-tert-butyl-6-(5-chloro-2H-benzotriazol-2-yl)phenol [CAS:3864-99-1] 2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol [CAS:25973-55-1] 2 1 * 1 mL
Catalog # ACSDRD1190551.2
Supplier:  CPACHEM