ContactUsLinkComponent
~MICROSLIDE 103x76,3±0,3 mm, thickness 1 mm, white glass, cut edges, without frosted end, paper-interleaved, in boxes of 50 pcs. MARIENFELD SUPERIOR 1 * 50 items
Catalog # MARI152310309
Supplier:  MARIENFELD
New Product
~MICROSLIDE 103x76,3±0,3 mm, thickness 1 mm, white glass, cut edges, without frosted end, paper-interleaved, in boxes of 50 pcs. MARIENFELD SUPERIOR 1 * 50 items
Catalog # MARI152310309
Supplier:  MARIENFELD