4,4-Difluorocyclohexanone, 97% 1 * 100 g
Catalog # ABCRAB205371-100G
Supplier: abcr
Frequently Bought Together
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Trifluoroacetic acid, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR®, Silicone Lab Mat
Supplier: VWR Collection