HSW AT 2P 50ML (60ML) LUER LOCK NOT FOR HUMAN USE 1 * 30 items
Catalog # HSWA8300065431
Supplier: HENKE SASS WOLF
New Product
HSW AT 2P 50ML (60ML) LUER LOCK NOT FOR HUMAN USE 1 * 30 items
Catalog # HSWA8300065431
Supplier: HENKE SASS WOLF
New Product
Frequently Bought Together
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Sodium hydroxide 30% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
Single-use syringes, 2-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF