ContactUsLinkComponent
12791/BS 718, 1.000-1.100:0.002g/cm , Accuracy +/- 1 scale division, 250mm long, reference temperature 20°C, without thermometer, bottom reading, calibrated, with DKD certificate (DAkkS) with 3 test points 1 * 1 items
Catalog # AMARH809608/DKD3
Supplier:  Amarell
New Product
12791/BS 718, 1.000-1.100:0.002g/cm , Accuracy +/- 1 scale division, 250mm long, reference temperature 20°C, without thermometer, bottom reading, calibrated, with DKD certificate (DAkkS) with 3 test points 1 * 1 items
Catalog # AMARH809608/DKD3
Supplier:  Amarell