Prostředek čistící Neodisher LaboClean A8 1 * 10 kg
WEIG461476
New Product
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
Pipette tips, Finntip™
: Thermo Fisher Scientific
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Hydrochloric acid 0.2 mol/l (0.2 N)
: Lach-ner S.R.O.