Prostředek kyselý neutr. Neodisher Z 1 * 10 L
Catalog # WEIG475230
Supplier: Dr. Weigert
New Product
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Prostředek čistící Neodisher LaboClean FLA 1 * 10 L
Supplier: Dr. Weigert