ContactUsLinkComponent
BakerBond Q2100 - C8 100 x 2.1mm 3µm 1 * 1 items
Catalog # 7354-12
Supplier:  Avantor
New Product
BakerBond Q2100 - C8 100 x 2.1mm 3µm 1 * 1 items
Catalog # 7354-12
Supplier:  Avantor