ContactUsLinkComponent
Leak-Sensor, Capacitive level sensor with mounting bracket made of PP for mounting in leakage trays in safety cabinets. Delivery includes two lock nuts for fastening the sensor in the bracket and setting the alarm time, 1 self-adhesive pad and 2 scre 1 * 1 items
Catalog # BOHLR637-01
Supplier:  Bohlender
New Product
Leak-Sensor, Capacitive level sensor with mounting bracket made of PP for mounting in leakage trays in safety cabinets. Delivery includes two lock nuts for fastening the sensor in the bracket and setting the alarm time, 1 self-adhesive pad and 2 scre 1 * 1 items
Catalog # BOHLR637-01
Supplier:  Bohlender