ContactUsLinkComponent
Desktop Mount Universal, PE-EX, connections top side: 1x male thread GL 45, 8 x capillary connections (UNF 1/4 inclined) and 4 connections for tubing ID 5,5-8,0 mm (NPT 1/8 straight). With self-adhesive, silicone-free sealing washer. Connection bot 1 * 1 items
Catalog # BOHLR1133-04
Supplier:  Bohlender
New Product
Desktop Mount Universal, PE-EX, connections top side: 1x male thread GL 45, 8 x capillary connections (UNF 1/4 inclined) and 4 connections for tubing ID 5,5-8,0 mm (NPT 1/8 straight). With self-adhesive, silicone-free sealing washer. Connection bot 1 * 1 items
Catalog # BOHLR1133-04
Supplier:  Bohlender