Powder funnel short, wide stem, PP out.dia. 80 mm stem dia. 23 mm l. 26 mm 1 * 10 items
BRND148025
: Brand
New Product
Powder funnel short, wide stem, PP out.dia. 80 mm stem dia. 23 mm l. 26 mm 1 * 10 items
BRND148025
: Brand
New Product
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR®, Safety Pipette Fillers, Standard/Universal/Flip
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Sodium hydroxide 97.0-100.5%, pellets NF, JP, Ph. Eur., BP
: VWR Chemicals
VWR®, Latex Gloves
: VWR Collection
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
: VWR Collection