9H-Fluoren-9-ylmethyl N-{[methoxy(methyl)carbamoyl]methyl}carbamate 1 * 5 g
Catalog # ABCRAB421340
Supplier: abcr
9H-Fluoren-9-ylmethyl N-{[methoxy(methyl)carbamoyl]methyl}carbamate 1 * 5 g
Catalog # ABCRAB421340
Supplier: abcr
Frequently Bought Together
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
TERT. BUTANOL FOR HPLC 99,5% 1 * 1 L
Supplier: Merck
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals